missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bis(diphenylphosphino)biphenyl 98.0+%, TCI America™

Catalog No. B2630100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2630100MG 100 mg
B26301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2630100MG Supplier TCI America Supplier No. B2630100MG

Chemical Identifiers

CAS 84783-64-2
Molecular Formula C36H28P2
Molecular Weight (g/mol) 522.57
MDL Number MFCD03094574
InChI Key GRTJBNJOHNTQBO-UHFFFAOYSA-N
Synonym 2,2'-bis diphenylphosphino-1,1'-biphenyl, 2,2'-bis diphenylphosphino biphenyl, biphep, 2-2-diphenylphosphanylphenyl phenyl-diphenyl-phosphane, dpbp, 2'-diphenylphosphanyl-1,1'-biphenyl-2-yl diphenylphosphane, pubchem9102, acmc-209pwq, 2,2'-bis diphenylphosphino-biphenyl, ar-2,2'-bis diphenylphosphino biphenyl
PubChem CID 2734940
IUPAC Name [2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 215°C
Color White-Yellow
Quantity 100 mg
Formula Weight 522.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bis(diphenylphosphino)biphenyl