missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(bromomethyl)-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7126941
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D180825G 25 g
D1808500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D180825G Supplier TCI America Supplier No. D180825G

Chemical Identifiers

CAS 3296-90-0
Molecular Formula C5H10Br2O2
Molecular Weight (g/mol) 261.94
MDL Number MFCD00004688
InChI Key CHUGKEQJSLOLHL-UHFFFAOYSA-N
Synonym Dibromoneopentyl Glycol
PubChem CID 18692
ChEBI CHEBI:82294
IUPAC Name 2,2-bis(bromomethyl)propane-1,3-diol
SMILES OCC(CO)(CBr)CBr

Specifications

Melting Point 111°C
Color White
Quantity 25 g
Formula Weight 261.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(bromomethyl)-1,3-propanediol
EINECSNumber (2)-2405
RTECSNumber TY3195500
TSCA Yes