accessibility menu, dialog, popup

UserName

OHHOBrBr

CAS RN 3296-90-0

IUPAC Name: 2,2-bis(bromomethyl)propane-1,3-diol
Synonyms: 2,2-bis(bromomethyl)propane-1,3-diol
Molecular Weight (g/mol): 261.94
Molecular Formula: C5H10Br2O2
InChi Key: CHUGKEQJSLOLHL-UHFFFAOYSA-N
SMILES: OCC(CO)(CBr)CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results