missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-hydroxyphenyl)propane 99.0+%, TCI America™
$46.60 - $46.60
Chemical Identifiers
| CAS | 80-05-7 |
|---|---|
| Molecular Formula | C15H16O2 |
| Molecular Weight (g/mol) | 228.29 |
| MDL Number | MFCD00002366 |
| InChI Key | IISBACLAFKSPIT-UHFFFAOYSA-N |
| Synonym | bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian |
| PubChem CID | 6623 |
| ChEBI | CHEBI:33216 |
| IUPAC Name | 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol |
| SMILES | CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 80-05-7 | |
| 228.29 | |
| IISBACLAFKSPIT-UHFFFAOYSA-N | |
| 6623 | |
| 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol |
| C15H16O2 | |
| MFCD00002366 | |
| bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian | |
| CHEBI:33216 | |
| CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 |
Specifications
| 80-05-7 | |
| White | |
| C15H16O2 | |
| 500 g | |
| IISBACLAFKSPIT-UHFFFAOYSA-N | |
| 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol | |
| 6623 | |
| 228.29 | |
| Crystalline Powder |
| 156°C | |
| 252°C | |
| MFCD00002366 | |
| bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian | |
| CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 | |
| 228.29 | |
| CHEBI:33216 | |
| ≥99.0% (GC) | |
| 2,2-Bis(4-hydroxyphenyl)propane |
Safety and Handling
EINECSNumber : (4)-0123
RTECSNumber : SL6300000
TSCA : Yes