missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-hydroxyphenyl)hexafluoropropane 98.0+%, TCI America™

Catalog No. p-7118762
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09455G 5 g
B094525G 25 g
B0945100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B09455G Supplier TCI America Supplier No. B09455G

Chemical Identifiers

CAS 1478-61-1
Molecular Formula C15H10F6O2
Molecular Weight (g/mol) 336.23
MDL Number MFCD00000439
InChI Key ZFVMWEVVKGLCIJ-UHFFFAOYSA-N
Synonym bisphenol af, 4,4'-hexafluoroisopropylidene diphenol, 2,2-bis 4-hydroxyphenyl hexafluoropropane, hexafluorobisphenol a, biphenol af, 4,4'-perfluoropropane-2,2-diyl diphenol, hexafluoroisopropylidenebis 4-hydroxybenzene, hexafluorodiphenylolpropane, hexafluoroacetone bisphenol a, 2,2-bis 4-hydroxyphenyl perfluoropropane
PubChem CID 73864
ChEBI CHEBI:72754
IUPAC Name 4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 161°C
Quantity 5 g
Formula Weight 336.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-hydroxyphenyl)hexafluoropropane
EINECSNumber (4)-1335
RTECSNumber SN2780000
TSCA Yes