missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-hydroxy-3-methylphenyl)propane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 139°C |
| Color | White |
| Quantity | 25 g |
| Synonym | 4,4′C-Isopropylidenedi-o-cresol |
| Formula Weight | 256.35 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2-Bis(4-hydroxy-3-methylphenyl)propane |
Safety and Handling
| EINECSNumber | (4)-1696 |
| RTECSNumber | GP1750000 |
| TSCA | Yes |