missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-hydroxy-3,5-dimethylphenyl)propane 98.0+%, TCI America™

Catalog No. B146425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B146425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B146425G Supplier TCI America Supplier No. B146425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5613-46-7
Molecular Formula C19H24O2
Molecular Weight (g/mol) 284.399
MDL Number MFCD00068238
InChI Key ODJUOZPKKHIEOZ-UHFFFAOYSA-N
Synonym 4,4′C-Isopropylidenebis(2,6-dimethylphenol), Tetramethylbisphenol A
PubChem CID 79717
IUPAC Name 4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)C(C)(C)C2=CC(=C(C(=C2)C)O)C

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 25 g
Formula Weight 284.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-hydroxy-3,5-dimethylphenyl)propane
EINECSNumber (4)-0214
TSCA Yes