missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-chloroformyloxyphenyl)propane 97.0+%, TCI America™
Supplier: TCI America B13351G
Specifications
| 2,2-Bis(4-chloroformyloxyphenyl)propane | |
| 94°C | |
| 216°C | |
| MFCD00060109 | |
| 2923 | |
| MMWCQWOKHLEYSP-UHFFFAOYSA-N | |
| 4-(2-{4-[(chlorocarbonyl)oxy]phenyl}propan-2-yl)phenyl carbonochloridate | |
| 74843 | |
| 353.20 | |
| Crystalline Powder |
| 2024-88-6 | |
| White-Yellow | |
| C17H14Cl2O4 | |
| 1 g | |
| bisphenol a bis chloroformate, 2,2-bis 4-chloroformyloxyphenyl propane, bisphenol a dichloroformate, carbonochloridic acid, 1-methylethylidene di-4,1-phenylene ester, 2,2-bis p-chloroformyloxy phenyl propane, carbonochloridic acid, c,c'-1-methylethylidene di-4,1-phenylene ester, isopropylidenedi-p-phenylene bis chloroformate, bisphenol a bischloroformate, bidd:er0407 | |
| CC(C)(C1=CC=C(OC(Cl)=O)C=C1)C1=CC=C(OC(Cl)=O)C=C1 | |
| 353.20 | |
| CHEBI:34576 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 2024-88-6 | |
| 353.20 | |
| MMWCQWOKHLEYSP-UHFFFAOYSA-N | |
| 74843 | |
| 4-(2-{4-[(chlorocarbonyl)oxy]phenyl}propan-2-yl)phenyl carbonochloridate |
| C17H14Cl2O4 | |
| MFCD00060109 | |
| bisphenol a bis chloroformate, 2,2-bis 4-chloroformyloxyphenyl propane, bisphenol a dichloroformate, carbonochloridic acid, 1-methylethylidene di-4,1-phenylene ester, 2,2-bis p-chloroformyloxy phenyl propane, carbonochloridic acid, c,c'-1-methylethylidene di-4,1-phenylene ester, isopropylidenedi-p-phenylene bis chloroformate, bisphenol a bischloroformate, bidd:er0407 | |
| CHEBI:34576 | |
| CC(C)(C1=CC=C(OC(Cl)=O)C=C1)C1=CC=C(OC(Cl)=O)C=C1 |
Safety and Handling
TSCA : Yes