missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(4-chloroformyloxyphenyl)propane 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 94°C |
| Color | White-Yellow |
| Boiling Point | 216°C |
| Quantity | 5 g |
| UN Number | 2923 |
| Formula Weight | 353.20 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2-Bis(4-chloroformyloxyphenyl)propane |
Safety and Handling
| TSCA | Yes |