missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-carboxyphenyl)hexafluoropropane 98.0+%, TCI America™

Catalog No. p-7119033
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14015G 5 g
B140125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14015G Supplier TCI America Supplier No. B14015G

Chemical Identifiers

CAS 1171-47-7
Molecular Formula C17H10F6O4
Molecular Weight (g/mol) 392.253
MDL Number MFCD00040931
InChI Key PHQYMDAUTAXXFZ-UHFFFAOYSA-N
Synonym 2,2-bis 4-carboxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dibenzoic acid, benzoic acid, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 4-2-4-carboxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl benzoic acid, 4,4'-hexafluoroisopropylidene bis benzoic acid, 4-1-4-carboxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethyl benzoic acid, acmc-1c1gj, 2,2-bis-4-carboxyphenyl hexafluoropropane
PubChem CID 633763
IUPAC Name 4-[2-(4-carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C(C2=CC=C(C=C2)C(=O)O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 275°C
Color White
Quantity 5 g
Formula Weight 392.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-carboxyphenyl)hexafluoropropane