missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-aminophenyl)hexafluoropropane 98.0+%, TCI America™

Catalog No. p-7119032
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14001G 1 g
B14005G 5 g
B140025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B14001G Supplier TCI America Supplier No. B14001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1095-78-9
Molecular Formula C15H12F6N2
Molecular Weight (g/mol) 334.265
MDL Number MFCD00039146
InChI Key BEKFRNOZJSYWKZ-UHFFFAOYSA-N
Synonym 2,2-bis 4-aminophenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dianiline, 4,4'-hexafluoroisopropylidene dianiline, 4-2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-yl aniline, benzenamine, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, acmc-1bon7, bekfrnozjsywkz-uhfffaoysa, 2,2-bis-4-aminophenyl hexafluoropropane, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl dianiline
PubChem CID 622236
IUPAC Name 4-[2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C(F)(F)F)C(F)(F)F)N

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 1 g
Formula Weight 334.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-aminophenyl)hexafluoropropane