missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[4-(4-aminophenoxy)phenyl]propane 98.0+%, TCI America™

Catalog No. B155125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B155125G 25 g
B1551250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B155125G Supplier TCI America Supplier No. B155125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13080-86-9
Molecular Formula C27H26N2O2
Molecular Weight (g/mol) 410.517
MDL Number MFCD00039152
InChI Key KMKWGXGSGPYISJ-UHFFFAOYSA-N
Synonym 2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, 2,2'-bis 4-aminophenoxyphenyl propane, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, bis 4-4-aminophenoxy phenyl dimethyl methane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine
PubChem CID 83119
IUPAC Name 4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline
SMILES CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N

Specifications

Melting Point 129°C
Quantity 25 g
Formula Weight 410.52
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis[4-(4-aminophenoxy)phenyl]propane
EINECSNumber (4)-1581
RTECSNumber CY1062500
TSCA Yes