missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(3-nitrophenyl)hexafluoropropane 99.0+%, TCI America™

Catalog No. b21151g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21151G 1 g
B21155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21151G Supplier TCI America Supplier No. B21151G

Chemical Identifiers

CAS 64465-34-5
Molecular Formula C15H8F6N2O4
Molecular Weight (g/mol) 394.23
MDL Number MFCD02093448
InChI Key YETXOTPYXAYEMI-UHFFFAOYSA-N
PubChem CID 14029053
IUPAC Name 1-[1,1,1,3,3,3-hexafluoro-2-(3-nitrophenyl)propan-2-yl]-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(=C1)C(C1=CC(=CC=C1)[N+]([O-])=O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 1 g
Synonym 3,3′C-(Hexafluoroisopropylidene)bis(nitrobenzene)
Formula Weight 394.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(3-nitrophenyl)hexafluoropropane