missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bis(2-oxazoline) 97.0+%, TCI America™
$190.13 - $562.95
Chemical Identifiers
| CAS | 36697-72-0 |
|---|---|
| Molecular Formula | C6H8N2O2 |
| Molecular Weight (g/mol) | 140.14 |
| MDL Number | MFCD00059930 |
| InChI Key | KKKKCPPTESQGQH-UHFFFAOYSA-N |
| PubChem CID | 283385 |
| IUPAC Name | 2-(4,5-dihydro-1,3-oxazol-2-yl)-4,5-dihydro-1,3-oxazole |
| SMILES | C1CN=C(O1)C1=NCCO1 |
Chemical Identifiers
| 36697-72-0 | |
| 140.14 | |
| KKKKCPPTESQGQH-UHFFFAOYSA-N | |
| 2-(4,5-dihydro-1,3-oxazol-2-yl)-4,5-dihydro-1,3-oxazole |
| C6H8N2O2 | |
| MFCD00059930 | |
| 283385 | |
| C1CN=C(O1)C1=NCCO1 |
Specifications
| 36697-72-0 | |
| C6H8N2O2 | |
| 5 g | |
| C1CN=C(O1)C1=NCCO1 | |
| 140.14 | |
| 140.14 | |
| Crystalline Powder |
| 213°C | |
| MFCD00059930 | |
| KKKKCPPTESQGQH-UHFFFAOYSA-N | |
| 2-(4,5-dihydro-1,3-oxazol-2-yl)-4,5-dihydro-1,3-oxazole | |
| 283385 | |
| ≥97.0% (N) | |
| 2,2′-Bis(2-oxazoline) |
Safety and Handling
EINECSNumber : (5)-5584
TSCA : No
Recommended Storage : Refrigerator