accessibility menu, dialog, popup

UserName

NONO

CAS RN 36697-72-0

IUPAC Name: 2-(4,5-dihydro-1,3-oxazol-2-yl)-4,5-dihydro-1,3-oxazole
Synonyms: 2-(4,5-dihydro-1,3-oxazol-2-yl)-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: KKKKCPPTESQGQH-UHFFFAOYSA-N
SMILES: C1CN=C(O1)C1=NCCO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results