missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine-6,6'-diol 98.0+%, TCI America™

$246.05 - $738.16

Chemical Identifiers

CAS 103505-54-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD13194839
InChI Key POJMWJLYXGXUNU-UHFFFAOYSA-N
PubChem CID 363601
IUPAC Name 6-(6-oxo-1H-pyridin-2-yl)-1H-pyridin-2-one
SMILES C1=CC(=O)NC(=C1)C2=CC=CC(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39661G
View Documents
TCI America
B39661G
1 g
Each for $246.05
Only null left
 
B39665G
View Documents
TCI America
B39665G
5 g
Each for $738.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

103505-54-0
188.186
POJMWJLYXGXUNU-UHFFFAOYSA-N
6-(6-oxo-1H-pyridin-2-yl)-1H-pyridin-2-one
C10H8N2O2
MFCD13194839
363601
C1=CC(=O)NC(=C1)C2=CC=CC(=O)N2

Specifications

103505-54-0
Yellow
MFCD13194839
6,6′C-Dihydroxy-2,2′C-bipyridine, 6,6′C-Dihydroxy-2,2′C-bipyridyl
C1=CC(=O)NC(=C1)C2=CC=CC(=O)N2
188.186
188.19
Crystalline Powder
340°C
C10H8N2O2
1 g
POJMWJLYXGXUNU-UHFFFAOYSA-N
6-(6-oxo-1H-pyridin-2-yl)-1H-pyridin-2-one
363601
≥98.0% (GC,T)
2,2′-Bipyridine-6,6′-diol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'