accessibility menu, dialog, popup

UserName

OHNONH

CAS RN 103505-54-0

IUPAC Name: 1H,1'H,6H,6'H-[2,2'-bipyridine]-6,6'-dione
Synonyms: 1H,1'H-[2,2'-bipyridine]-6,6'-dione
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: POJMWJLYXGXUNU-UHFFFAOYSA-N
SMILES: O=C1NC(=CC=C1)C1=CC=CC(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results