missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bipyridine-6,6'-diol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 340°C |
| Color | Yellow |
| Quantity | 5 g |
| Synonym | 6,6′-Dihydroxy-2,2′-bipyridine, 6,6′-Dihydroxy-2,2′-bipyridyl |
| Formula Weight | 188.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Bipyridine-6,6′-diol |
Safety and Handling
| TSCA | No |