missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biimidazole

Catalog No. AC465820010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465820010 1 g
AC465820050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465820010 Supplier Thermo Scientific Chemicals Supplier No. 465820010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 492-98-8 | C6H6N4 | 134.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-98-8
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00047014
InChI Key AZUHIVLOSAPWDM-UHFFFAOYSA-N
Synonym 2,2 '-Bisimidazole; 2,2 '-Diimidazole
IUPAC Name 1H,1'H-2,2'-biimidazole
SMILES N1C=CN=C1C1=NC=CN1

Specifications

Alpha Vector BIIMIDAZOLE
Melting Point >350°C
Color Gray to Yellow
Infrared Spectrum Conforms
Assay Percent Range 97.5 to 102.5 % (Titration with HClO4)
Packaging glass bottle
Quantity 1 g
UN Number 3265
Formula Weight 134.14
Physical Form Crystalline Powder
Chemical Name or Material 2,2 '-Biimidazole
Show More Show Less
Hazard Category GHS05: Corrosive
Hazard Statement GHS07: Harmful
Precautionary Statement GHS08: Health hazard
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only