missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Biimidazole 98.0+%, TCI America™
$262.83 - $911.52
Chemical Identifiers
| CAS | 492-98-8 |
|---|---|
| Molecular Formula | C6H6N4 |
| Molecular Weight (g/mol) | 134.14 |
| MDL Number | MFCD00047014 |
| InChI Key | AZUHIVLOSAPWDM-UHFFFAOYSA-N |
| PubChem CID | 101463 |
| IUPAC Name | 1H,1'H-2,2'-biimidazole |
| SMILES | N1C=CN=C1C1=NC=CN1 |
Chemical Identifiers
| 492-98-8 | |
| 134.14 | |
| AZUHIVLOSAPWDM-UHFFFAOYSA-N | |
| 1H,1'H-2,2'-biimidazole |
| C6H6N4 | |
| MFCD00047014 | |
| 101463 | |
| N1C=CN=C1C1=NC=CN1 |
Specifications
| 492-98-8 | |
| C6H6N4 | |
| 1 g | |
| N1C=CN=C1C1=NC=CN1 | |
| 134.14 | |
| 134.14 | |
| Crystalline Powder |
| 300°C | |
| MFCD00047014 | |
| AZUHIVLOSAPWDM-UHFFFAOYSA-N | |
| 1H,1'H-2,2'-biimidazole | |
| 101463 | |
| ≥98.0% (HPLC,T) | |
| 2,2′-Biimidazole |
Safety and Handling
TSCA : No