missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biimidazole 98.0+%, TCI America™

Catalog No. B24871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24871G 1 g
B24875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24871G Supplier TCI America Supplier No. B24871G

Chemical Identifiers

CAS 492-98-8
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00047014
InChI Key AZUHIVLOSAPWDM-UHFFFAOYSA-N
PubChem CID 101463
IUPAC Name 1H,1'H-2,2'-biimidazole
SMILES N1C=CN=C1C1=NC=CN1

Specifications

Melting Point 300°C
Quantity 1 g
Formula Weight 134.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Biimidazole