missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bicinchoninic Acid Dipotassium Salt Hydrate 98.0+%, TCI America™
Supplier: TCI America B10775G
Specifications
| 2,2′-Bicinchoninic Acid Dipotassium Salt Hydrate [for Determination of Cu] | |
| C20H12K2N2O4 | |
| 5 g | |
| HRNSKWTYGVQLDN-UHFFFAOYSA-N | |
| 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium | |
| 66596657 | |
| ≥98.0% (HPLC) |
| 63451-34-3 | |
| MFCD00014664 | |
| 2,2′-Biquinoline-4,4′-dicarboxylic Acid Dipotassium Salt, 4,4′-Dicarboxy-2,2′-biquinoline Dipotassium Salt | |
| C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K] | |
| 422.523 | |
| 420.51 | |
| Crystalline Powder |
Chemical Identifiers
| 63451-34-3 | |
| 422.523 | |
| HRNSKWTYGVQLDN-UHFFFAOYSA-N | |
| 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium |
| C20H12K2N2O4 | |
| MFCD00014664 | |
| 66596657 | |
| C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K] |
Safety and Handling
TSCA : Yes