CAS RN 63451-34-3
CAS RN 63451-34-3
IUPAC Name:
dipotassium [2,2'-biquinoline]-4,4'-dicarboxylate
Synonyms:
dipotassium [2,2'-biquinoline]-4,4'-dicarboxylate
Molecular Weight (g/mol):
420.51
Molecular Formula:
C20H10K2N2O4
InChi Key:
WCYSOCFXPUNTJG-UHFFFAOYSA-L
SMILES:
[K+].[K+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
1
–
1
of
1
results
1
2,2'-Bicinchoninic Acid Dipotassium Salt Hydrate 98.0+%, TCI America™
CAS: 63451-34-3 Molecular Formula: C20H12K2N2O4 Molecular Weight (g/mol): 422.523 MDL Number: MFCD00014664 InChI Key: HRNSKWTYGVQLDN-UHFFFAOYSA-N PubChem CID: 66596657 IUPAC Name: 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium SMILES: C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K]