missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bicinchoninic Acid Dipotassium Salt Hydrate 98.0+%, TCI America™

Catalog No. B10771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10771G 1 g
B10775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10771G Supplier TCI America Supplier No. B10771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63451-34-3
Molecular Formula C20H12K2N2O4
Molecular Weight (g/mol) 422.523
MDL Number MFCD00014664
InChI Key HRNSKWTYGVQLDN-UHFFFAOYSA-N
PubChem CID 66596657
IUPAC Name 2-(4-carboxyquinolin-2-yl)quinoline-4-carboxylic acid;potassium
SMILES C1=CC=C2C(=C1)C(=CC(=N2)C3=NC4=CC=CC=C4C(=C3)C(=O)O)C(=O)O.[K].[K]

Specifications

Quantity 1 g
Synonym 2,2′C-Biquinoline-4,4′C-dicarboxylic Acid Dipotassium Salt, 4,4′C-Dicarboxy-2,2′C-biquinoline Dipotassium Salt
Formula Weight 420.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bicinchoninic Acid Dipotassium Salt Hydrate [for Determination of Cu]