missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Benzidinedisulfonic Acid (contains 30% Water at maximum) 70.0+%, TCI America™

Catalog No. p-7118421
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B039525G 25 g
B0395500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B039525G Supplier TCI America Supplier No. B039525G

Chemical Identifiers

CAS 117-61-3
Molecular Formula C12H12N2O6S2
Molecular Weight (g/mol) 344.36
MDL Number MFCD00041885
InChI Key MBJAPGAZEWPEFB-UHFFFAOYSA-N
Synonym 2,2'-benzidinedisulfonic acid, 2,2'-disulfobenzidine, benzidine-2,2'-disulfonic acid, 6,6'-bimetanilic acid, 4,4'-diaminobiphenyl-2,2'-disulfonic acid, benzidine-2,2'-disulphonic acid, 4,4'-diamino-1,1'-biphenyl-2,2'-disulfonic acid, unii-alf643n2eq, benzidine, 2,2'-disulfo, 4,4'-diamino-2,2'-biphenyldisulfonic acid
PubChem CID 8337
IUPAC Name 4,4'-diamino-[1,1'-biphenyl]-2,2'-disulfonic acid
SMILES NC1=CC=C(C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O

Specifications

Quantity 25 g
Formula Weight 344.36
Percent Purity ≥70.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Benzidinedisulfonic Acid (contains 30% Water at maximum)
EINECSNumber (4)-0836
RTECSNumber DV5230000
TSCA Yes