missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America A3012100G
Specifications
| 2,2′-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride | |
| 190°C | |
| C12H24Cl2N6 | |
| 100 g | |
| [H+].[H+].[Cl-].[Cl-].CC(CN=NCC(C)C1=NCCN1)C1=NCCN1 | |
| 323.27 | |
| 323.27 | |
| Crystalline Powder |
| 27776-21-2 | |
| White-Yellow | |
| MFCD00142723 | |
| ZVVWZIADMMTPBX-UHFFFAOYNA-N | |
| dihydrogen 2-(1-{2-[2-(4,5-dihydro-1H-imidazol-2-yl)propyl]diazen-1-yl}propan-2-yl)-4,5-dihydro-1H-imidazole dichloride | |
| 9949101 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 27776-21-2 | |
| 323.27 | |
| ZVVWZIADMMTPBX-UHFFFAOYNA-N | |
| dihydrogen 2-(1-{2-[2-(4,5-dihydro-1H-imidazol-2-yl)propyl]diazen-1-yl}propan-2-yl)-4,5-dihydro-1H-imidazole dichloride |
| C12H24Cl2N6 | |
| MFCD00142723 | |
| 9949101 | |
| [H+].[H+].[Cl-].[Cl-].CC(CN=NCC(C)C1=NCCN1)C1=NCCN1 |
Safety and Handling
RTECSNumber : NI3506608
TSCA : Yes