missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7118194
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A301225G 25 g
A3012100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A301225G Supplier TCI America Supplier No. A301225G
Only null left

Chemical Identifiers

CAS 27776-21-2
Molecular Formula C12H24Cl2N6
Molecular Weight (g/mol) 323.27
MDL Number MFCD00142723
InChI Key ZVVWZIADMMTPBX-UHFFFAOYNA-N
PubChem CID 9949101
IUPAC Name dihydrogen 2-(1-{2-[2-(4,5-dihydro-1H-imidazol-2-yl)propyl]diazen-1-yl}propan-2-yl)-4,5-dihydro-1H-imidazole dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CC(CN=NCC(C)C1=NCCN1)C1=NCCN1

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 25 g
Formula Weight 323.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride
RTECSNumber NI3506608
TSCA Yes