missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminophenyl)benzimidazole, 98%

Catalog No. AAL0703406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0703406 5 g
AAL0703414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0703406 Supplier Thermo Scientific Chemicals Supplier No. L0703406

CAS: 5805-39-0 | C13H11N3 | 209.25 g/mol

2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5805-39-0
Molecular Formula C13H11N3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00051496
InChI Key YWNXHTNWOQHFRL-UHFFFAOYSA-N
Synonym 2-2-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl aniline, 2-1h-benzimidazol-2-yl aniline, 2-1h-benzoimidazol-2-yl-phenylamine, 2-1h-1,3-benzodiazol-2-yl aniline, benzenamine, 2-1h-benzimidazol-2-yl, 2-o-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl benzenamine, 2-benzimidazol-2-ylphenylamine
PubChem CID 79869
SMILES NC1=CC=CC=C1C1=NC2=CC=CC=C2N1

Specifications

Melting Point 212°C to 216°C
Quantity 5 g
Beilstein 173018
Solubility Information Insoluble in water.
IUPAC Name 2-(1H-1,3-benzodiazol-2-yl)aniline
Formula Weight 209.24
Percent Purity 98%
Chemical Name or Material 2-(2-Aminophenyl)benzimidazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 227-361-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only