CAS RN 5805-39-0
IUPAC Name:
2-(1H-1,3-benzodiazol-2-yl)aniline
Synonyms:
2-(1H-1,3-benzodiazol-2-yl)aniline
Molecular Weight (g/mol):
209.25
Molecular Formula:
C13H11N3
InChi Key:
YWNXHTNWOQHFRL-UHFFFAOYSA-N
SMILES:
NC1=CC=CC=C1C1=NC2=CC=CC=C2N1
1
–
1
of
1
results
1
Promotions Available
2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
CAS: 5805-39-0 Molecular Formula: C13H11N3 Molecular Weight (g/mol): 209.25 MDL Number: MFCD00051496 InChI Key: YWNXHTNWOQHFRL-UHFFFAOYSA-N Synonym: 2-2-aminophenyl benzimidazole,2-1h-benzo d imidazol-2-yl aniline,2-1h-benzimidazol-2-yl aniline,2-1h-benzoimidazol-2-yl-phenylamine,2-1h-1,3-benzodiazol-2-yl aniline,benzenamine, 2-1h-benzimidazol-2-yl,2-o-aminophenyl benzimidazole,2-1h-benzo d imidazol-2-yl benzenamine,2-benzimidazol-2-ylphenylamine PubChem CID: 79869 SMILES: NC1=CC=CC=C1C1=NC2=CC=CC=C2N1