missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethylamino)ethanol 99.0+%, TCI America™

Catalog No. A029925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A029925ML 25 mL
A0299500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A029925ML Supplier TCI America Supplier No. A029925ML

Chemical Identifiers

CAS 111-41-1
Molecular Formula C4H12N2O
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008170
InChI Key LHIJANUOQQMGNT-UHFFFAOYSA-N
Synonym 2-2-aminoethylamino ethanol, n-2-hydroxyethyl ethylenediamine, aminoethylethanolamine, n-2-aminoethyl ethanolamine, ethanol, 2-2-aminoethyl amino, 2-2-aminoethyl amino ethanol, monoethanolethylenediamine, n-hydroxyethyl ethylenediamine, n-aminoethyl ethanolamine, 2-aminoethyl ethanolamine
PubChem CID 8112
IUPAC Name 2-[(2-aminoethyl)amino]ethan-1-ol
SMILES NCCNCCO

Specifications

Color Yellow
Boiling Point 244°C
Quantity 25 mL
UN Number 2735
Formula Weight 104.15
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethylamino)ethanol
EINECSNumber (2)-0304
RTECSNumber KJ6300000
TSCA Yes