accessibility menu, dialog, popup

UserName

H2NNHOH

CAS RN 111-41-1

IUPAC Name: 2-[(2-aminoethyl)amino]ethan-1-ol
Synonyms: aminoethylethanolamine (aeea)
Molecular Weight (g/mol): 104.15
Molecular Formula: C4H12N2O
InChi Key: LHIJANUOQQMGNT-UHFFFAOYSA-N
SMILES: NCCNCCO
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)