missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Aminoethyl)pyridine 98.0+%, TCI America™
Supplier: TCI America A199925G
Specifications
| 2-(2-Aminoethyl)pyridine | |
| Yellow | |
| C7H10N2 | |
| 25 g | |
| 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine | |
| NCCC1=CC=CC=N1 | |
| 122.17 | |
| CHEBI:74024 | |
| ≥98.0% (GC,T) |
| 2706-56-1 | |
| 93°C | |
| MFCD00006367 | |
| 2735 | |
| XPQIPUZPSLAZDV-UHFFFAOYSA-N | |
| 2-(pyridin-2-yl)ethan-1-amine | |
| 75919 | |
| 122.17 | |
| Liquid |
Chemical Identifiers
| 2706-56-1 | |
| 122.17 | |
| XPQIPUZPSLAZDV-UHFFFAOYSA-N | |
| 75919 | |
| 2-(pyridin-2-yl)ethan-1-amine |
| C7H10N2 | |
| MFCD00006367 | |
| 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine | |
| CHEBI:74024 | |
| NCCC1=CC=CC=N1 |
Safety and Handling
TSCA : Yes