missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)pyridine 98.0+%, TCI America™

Catalog No. p-7117678
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19995G 5 g
A199925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19995G Supplier TCI America Supplier No. A19995G
Only null left

Chemical Identifiers

CAS 2706-56-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006367
InChI Key XPQIPUZPSLAZDV-UHFFFAOYSA-N
Synonym 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine
PubChem CID 75919
ChEBI CHEBI:74024
IUPAC Name 2-(pyridin-2-yl)ethan-1-amine
SMILES NCCC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 2735
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethyl)pyridine