missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethylcyclohexanone, 97%

CAS: 2408-37-9 | C9H16O | 140.226 g/mol

Supplier:  Thermo Scientific Chemicals B2111114

 View more versions of this product

Catalog No. AAB2111114


Only null left
Explore available promotions

Description

Description

2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Specifications

Specifications

2,2,6-Trimethylcyclohexanone
0.904
51°C (125°F)
1.447
25 g
1905593
Slightly miscible with water.
CC1CCCC(C1=O)(C)C
140.226
140.23
2408-37-9
178°C to 179°C
C9H16O
MFCD00045550
UN1224
2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s
ZPVOLGVTNLDBFI-UHFFFAOYSA-N
2,2,6-trimethylcyclohexan-1-one
17000
97%

Chemical Identifiers

2408-37-9
140.226
ZPVOLGVTNLDBFI-UHFFFAOYSA-N
17000
CC1CCCC(C1=O)(C)C
C9H16O
MFCD00045550
2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s
2,2,6-trimethylcyclohexan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 219-309-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only