missing translation for 'onlineSavingsMsg'
Learn More

2,2,6-Trimethylcyclohexanone, 97%

Catalog No. p-7025565
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2111103 1 g
AAB2111106 5 g
AAB2111114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2111103 Supplier Thermo Scientific Chemicals Supplier No. B2111103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2408-37-9 | C9H16O | 140.226 g/mol

2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6-Trimethylcyclohexanone is used as a flavoring agent. It is also used to prepare terpene viz. beta-ionone.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 2408-37-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00045550
InChI Key ZPVOLGVTNLDBFI-UHFFFAOYSA-N
Synonym 2,2,6-trimethylcyclohexanone, cyclohexanone, 2,2,6-trimethyl, 2,6,6-trimethylcyclohexanone, 1,1,3-trimethyl-2-cyclohexanone, +/--2,2,6-trimethylcyclohexanone, fema no. 3473, 2,6,6-trimethylcyclohexan-1-one, cistus cyclohexanone, cyclohexanone, 2,2,6-trimethyl-, r, cyclohexanone, 2,2,6-trimethyl-, s
PubChem CID 17000
IUPAC Name 2,2,6-trimethylcyclohexan-1-one
SMILES CC1CCCC(C1=O)(C)C

Specifications

Density 0.904
Boiling Point 178°C to 179°C
Flash Point 51°C (125°F)
Refractive Index 1.447
Quantity 1 g
UN Number UN1224
Beilstein 1905593
Solubility Information Slightly miscible with water.
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material 2,2,6-Trimethylcyclohexanone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 219-309-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only