missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dichlorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7127566
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29261G 1 g
D29265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29261G Supplier TCI America Supplier No. D29261G

Chemical Identifiers

CAS 14573-23-0
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.067
MDL Number MFCD00060612
InChI Key ACIMQXSSGMWVKG-UHFFFAOYSA-N
Synonym 2,6-dichlorophenethylamine, 2-2,6-dichlorophenyl ethanamine, 2-2,6-dichlorophenyl ethylamine, 2-2,6-dichlorophenyl ethan-1-amine, benzeneethanamine, 2,6-dichloro, 2-2,6-dichloro-phenyl-ethylamine, 2,6-dichlorophenylethylamine, pubchem12520, acmc-209cvj, ksc495o0f
PubChem CID 2734101
IUPAC Name 2-(2,6-dichlorophenyl)ethanamine
SMILES C1=CC(=C(C(=C1)Cl)CCN)Cl

Specifications

Color Yellow
Boiling Point 129°C
Quantity 1 g
Formula Weight 190.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2,6-Dichlorophenyl)ethylamine