accessibility menu, dialog, popup

UserName

NH3ClCl

CAS RN 14573-23-0

IUPAC Name: 2-(2,6-dichlorophenyl)ethan-1-aminium
Synonyms: 2-(2,6-dichlorophenyl)ethanaminium
Molecular Weight (g/mol): 191.07
Molecular Formula: C8H10Cl2N
InChi Key: ACIMQXSSGMWVKG-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results