missing translation for 'onlineSavingsMsg'
Learn More

2-(2,6-Dichloroanilino)-2-imidazoline Hydrochloride 98.0+%, TCI America™

Catalog No. D13531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13531G 1 g
D13535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13531G Supplier TCI America Supplier No. D13531G

Chemical Identifiers

CAS 4205-91-8
Molecular Formula C9H10Cl3N3
Molecular Weight (g/mol) 266.55
MDL Number MFCD00036705
InChI Key ZNIFSRGNXRYGHF-UHFFFAOYSA-N
Synonym clonidine hydrochloride, clonidine hcl, catapres, 2-2,6-dichloroanilino-2-imidazoline hydrochloride, chlophazolin, dispaclonidin, normopresan, atensina, barclyd, capresin
PubChem CID 20179
IUPAC Name N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine;hydrochloride
SMILES C1CN=C(N1)NC2=C(C=CC=C2Cl)Cl.Cl

Specifications

Melting Point 312°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 266.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,6-Dichloroanilino)-2-imidazoline Hydrochloride
EINECSNumber (9)-2186
RTECSNumber NJ2490000
TSCA No