accessibility menu, dialog, popup

UserName

HClClClNHNHN

CAS RN 4205-91-8

IUPAC Name: hydrogen N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine chloride
Synonyms: hydrogen clonidine chloride
Molecular Weight (g/mol): 266.55
Molecular Formula: C9H10Cl3N3
InChi Key: ZNIFSRGNXRYGHF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=CC(Cl)=C1NC1=NCCN1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)