missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidinooxy, 98%

CAS: 2564-83-2 | C9H18NO | 156.25 g/mol

$94.88 - $94.88

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9H18NO
Molecular Weight (g/mol) 156.25
MDL Number MFCD00009599
InChI Key QYTDEUPAUMOIOP-UHFFFAOYSA-N
Synonym TEMPO
IUPAC Name (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
SMILES CC1(C)CCCC(C)(C)N1[O]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC200380050
SDS View Documents
Thermo Scientific Chemicals
200380050
5 g Glass bottle
Each for $94.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

2564-83-2
156.25
QYTDEUPAUMOIOP-UHFFFAOYSA-N
(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
C9H18NO
MFCD00009599
TEMPO
CC1(C)CCCC(C)(C)N1[O]

Specifications

2564-83-2
100.0
Yellow to Green
Authentic
Glass bottle
MFCD00009599
16,324; 17,332
TEMPO
QYTDEUPAUMOIOP-UHFFFAOYSA-N
(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
156.25
Powder
97.5
36°C to 40°C
67°C
97.5 % min. (GC)
C9H18NO
5 g
15, 9283
Solubility in water: 151mg/l (20°C)
CC1(C)CCCC(C)(C)N1[O]
156.25
98%
2,2,6,6-Tetramethylpiperidinooxy, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 219-888-8

RTECSNumber : TN8991900

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only