CAS RN 2564-83-2
IUPAC Name:
(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms:
tanan tanane tetramethylpiperidine nitroxide oxoammonium tpo tempo tetramethylpiperidinooxy
Molecular Weight (g/mol):
156.25
Molecular Formula:
C9H18NO
InChi Key:
QYTDEUPAUMOIOP-UHFFFAOYSA-N
SMILES:
CC1(C)CCCC(C)(C)N1[O]
1
–
1
of
1
results
1
Promotions Available
TEMPO, free radical, 98+%
TEMPO, free radical, 98+%
CAS: 2564-83-2 Molecular Formula: C9H18NO Molecular Weight (g/mol): 156.25 MDL Number: MFCD00009599 InChI Key: QYTDEUPAUMOIOP-UHFFFAOYSA-N Synonym: 2,2,6,6-Tetramethyl-1-piperidinyloxy, free radical IUPAC Name: (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl SMILES: CC1(C)CCCC(C)(C)N1[O]