missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidine 98.0+%, TCI America™

Catalog No. p-7136575
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T105125ML 25 mL
T1051250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T105125ML Supplier TCI America Supplier No. T105125ML
Only null left

Chemical Identifiers

CAS 768-66-1
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.258
MDL Number MFCD00005985
InChI Key RKMGAJGJIURJSJ-UHFFFAOYSA-N
Synonym norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i
PubChem CID 13035
IUPAC Name 2,2,6,6-tetramethylpiperidine
SMILES CC1(CCCC(N1)(C)C)C

Specifications

Quantity 25 mL
UN Number 1992
Formula Weight 141.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine
RTECSNumber TN4220000
TSCA Yes