accessibility menu, dialog, popup

UserName

CAS RN 768-66-1

IUPAC Name: 2,2,6,6-tetramethylpiperidine
Synonyms: 2,2,6,6-tetramethylpiperidine
Molecular Weight (g/mol): 141.26
Molecular Formula: C9H19N
InChi Key: RKMGAJGJIURJSJ-UHFFFAOYSA-N
SMILES: CC1(C)CCCC(C)(C)N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)