missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidine, 98%

Catalog No. p-4510228
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151950100 10 g Glass bottle
AC151950250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151950100 Supplier Thermo Scientific Chemicals Supplier No. 151950100

CAS: 768-66-1 | C9H19N | 141.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-66-1
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.26
InChI Key RKMGAJGJIURJSJ-UHFFFAOYSA-N
Synonym norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i
PubChem CID 13035
IUPAC Name 2,2,6,6-tetramethylpiperidine
SMILES CC1(CCCC(N1)(C)C)C

Specifications

Color Colorless to Yellow
Density 0.8300g/mL
Boiling Point 152.0°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4460
Quantity 10 g
Beilstein 20,129
Specific Gravity 0.83
Solubility Information Solubility in water: miscible. Other solubilities: soluble in toluene, dimethylformamide
Formula Weight 141.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective glo
EINECSNumber 212-199-3

RUO – Research Use Only