missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. p-7136839
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15605G 5 g
T156025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15605G Supplier TCI America Supplier No. T15605G
Only null left

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9H18NO
Molecular Weight (g/mol) 156.25
MDL Number MFCD00009599
InChI Key QYTDEUPAUMOIOP-UHFFFAOYSA-N
Synonym TEMPO
IUPAC Name (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
SMILES CC1(C)CCCC(C)(C)N1[O]

Specifications

Melting Point 39°C
Quantity 5 g
UN Number 3263
Formula Weight 156.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine 1-Oxyl Free Radical
RTECSNumber TN8991900
TSCA Yes
Recommended Storage Refrigerator