missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-Oxyl Free Radical 98.0+%, TCI America™

Catalog No. p-7137618
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31691G 1 g
T31695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31691G Supplier TCI America Supplier No. T31691G
Only null left

Chemical Identifiers

CAS 147045-24-7
Molecular Formula C12H20NO2
Molecular Weight (g/mol) 210.297
InChI Key POEBHISKBXRUBH-UHFFFAOYSA-N
Synonym 2,2,6,6-Tetramethyl-4-(propargyloxy)piperidine 1-Oxyl, 4-(2-Propynyloxy)-TEMPO, 4-Propargyloxy-TEMPO
PubChem CID 59188306
IUPAC Name 1-$l^{1}-oxidanyl-2,2,6,6-tetramethyl-4-prop-2-ynoxypiperidine
SMILES CC1(CC(CC(N1[O])(C)C)OCC#C)C

Specifications

Melting Point 67°C
Quantity 1 g
Formula Weight 210.30
Percent Purity ≥98.0% (GC,T)
Physical Form Powder
Chemical Name or Material 2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-Oxyl Free Radical