accessibility menu, dialog, popup

UserName

CAS RN 147045-24-7

H3CNOH3CH3COCHCH3

CAS RN 147045-24-7

IUPAC Name: [2,2,6,6-tetramethyl-4-(prop-2-yn-1-yloxy)piperidin-1-yl]oxidanyl
Synonyms: 2,2,6,6-tetramethyl-4-(prop-2-yn-1-yloxy)piperidin-1-yloxidanyl
Molecular Weight (g/mol): 210.3
Molecular Formula: C12H20NO2
InChi Key: POEBHISKBXRUBH-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1[O])OCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results