missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-Oxyl Free Radical 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 67°C |
| Quantity | 5 g |
| Formula Weight | 210.30 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Powder |
| Chemical Name or Material | 2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-Oxyl Free Radical |
Safety and Handling
| TSCA | No |