missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2':6',2″-Terpyridine 98.0+%, TCI America™
Supplier: TCI America T00241G
Specifications
| 2,2′:6′,2′′-Terpyridine | |
| 92°C | |
| 370°C | |
| MFCD00006213 | |
| 2811 | |
| DRGAZIDRYFYHIJ-UHFFFAOYSA-N | |
| 6-(pyridin-2-yl)-2,2'-bipyridine | |
| 70848 | |
| 233.27 | |
| Crystalline Powder |
| 1148-79-4 | |
| Green-White | |
| C15H11N3 | |
| 1 g | |
| 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine | |
| C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1 | |
| 233.27 | |
| CHEBI:245199 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1148-79-4 | |
| 233.27 | |
| DRGAZIDRYFYHIJ-UHFFFAOYSA-N | |
| 70848 | |
| 6-(pyridin-2-yl)-2,2'-bipyridine |
| C15H11N3 | |
| MFCD00006213 | |
| 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine | |
| CHEBI:245199 | |
| C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1 |
Safety and Handling
TSCA : Yes