missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine, 97%

Catalog No. AAA1775001
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1775001 0.25 g
AAA1775003 1 g
AAA1775006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1775001 Supplier Thermo Scientific Chemicals Supplier No. A1775001
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1148-79-4 | C15H11N3 | 233.27 g/mol

2,2';6',2'-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′;6′,2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.

Solubility
Soluble in most organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1148-79-4
Molecular Formula C15H11N3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00006213
InChI Key DRGAZIDRYFYHIJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine
PubChem CID 70848
ChEBI CHEBI:245199
IUPAC Name 2,6-dipyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1

Specifications

Melting Point 89°C to 92°C
Odor Odorless
Assay Percent Range 0.97
Quantity 0.25 g
UN Number UN2811
Solubility Information Soluble in most organic solvents.
Formula Weight 233.28
Percent Purity 97%
Chemical Name or Material 2,2':6',2''-Terpyridine
Hazard Category H300+H310-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 214-559-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only